missing translation for 'onlineSavingsMsg'
Learn More

Boron carbide

Catalog No. AA4050430
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4050430 250 g
AA4050418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4050430 Supplier Thermo Scientific Chemicals Supplier No. 04050430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12069-32-8 | CB4 | 55.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12069-32-8
Molecular Formula CB4
Molecular Weight (g/mol) 55.25
MDL Number MFCD00011520
InChI Key INAHAJYZKVIDIZ-UHFFFAOYSA-N
Synonym boron carbide, boron carbide b4c, tetra 10b boron carbide, boron carbide, dust, tetrabor, boron carbide powder, boron carbide b4c grade hp, boron carbide b4c grade hs, b4-c, boron carbide b4c grade hd 07
PubChem CID 123279
IUPAC Name 2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane
SMILES B12B3B4B1C234

Specifications

Melting Point 2350°C
Physical Form Powder
Quantity 250 g
Merck Index 14,1344
Formula Weight 55.26
Odor Odorless
Density 2.52 g/mL
Chemical Name or Material Boron carbide
Hazard Category H315-H319-H332-H335
Hazard Statement GHS H Statement
H332-H315-H319-H335
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 235-111-5
RTECSNumber ED7420000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only