Learn More
Boron carbide
Description
Boron carbide is a hard boron-carbon ceramic material used in tank armor, bulletproof vests, engine sabotage powders, neutron absorber, cutting tools and dies and in brake linings of vehicles. It acts as an antioxidant additives in magnesia-carbon bricks. It is used as a precursor in the production of boron containing materials such as titanium boride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2350°C |
| Color | Black |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | >99% |
| Solubility Information | Insoluble in water. |
| Formula Weight | 55.26 |
| Odor | Odorless |
| Density | 2.52 g/cm3 |
| Chemical Name or Material | Boron carbide |
Safety and Handling
| Hazard Category | H315-H319-H332-H335 |
| Hazard Statement | Keep container in a well ventilated place. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 235-111-5 |
| RTECSNumber | ED7420000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only