Learn More
Boron carbide, 90+%
CAS: 12069-32-8 | CB4 | 55.25 g/mol
Supplier: Thermo Scientific Chemicals 01092222
Description
Boron carbide is a hard boron-carbon ceramic material used in tank armor, bulletproof vests, engine sabotage powders, neutron absorber, cutting tools and dies and in brake linings of vehicles. It acts as an antioxidant additives in magnesia-carbon bricks. It is used as a precursor in the production of boron containing materials such as titanium boride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Boron carbide | |
| 12069-32-8 | |
| Powder | |
| >90% | |
| MFCD00011520 | |
| Insoluble in water. | |
| B12B3B4B1C234 | |
| 55.25 | |
| 55.26 | |
| 2.52 |
| 2350°C | |
| Black | |
| 100 g | |
| CB4 | |
| boron carbide, boron carbide b4c, tetra 10b boron carbide, boron carbide, dust, tetrabor, boron carbide powder, boron carbide b4c grade hp, boron carbide b4c grade hs, b4-c, boron carbide b4c grade hd 07 | |
| INAHAJYZKVIDIZ-UHFFFAOYSA-N | |
| 2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,âµ]pentane | |
| 123279 | |
| Odorless |
Chemical Identifiers
| 12069-32-8 | |
| 55.25 | |
| INAHAJYZKVIDIZ-UHFFFAOYSA-N | |
| 123279 | |
| B12B3B4B1C234 |
| CB4 | |
| MFCD00011520 | |
| boron carbide, boron carbide b4c, tetra 10b boron carbide, boron carbide, dust, tetrabor, boron carbide powder, boron carbide b4c grade hp, boron carbide b4c grade hs, b4-c, boron carbide b4c grade hd 07 | |
| 2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,âµ]pentane |
Safety and Handling
Keep container in a well ventilated place. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H332-H335
EINECSNumber : 235-111-5
RTECSNumber : ED7420000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only