missing translation for 'onlineSavingsMsg'
Learn More

Bornyl Acetate (contains ca. 20% Isobornyl Acetate) 70.0+%, TCI America™

Catalog No. B052625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B052625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B052625ML Supplier TCI America Supplier No. B052625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5655-61-8
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00867808,MFCD00135942,MFCD00135942
InChI Key KGEKLUUHTZCSIP-HOSYDEDBSA-N
Synonym bornyl acetate, -bornyl acetate
PubChem CID 44630108
IUPAC Name (1S,2R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl acetate
SMILES CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 196.29
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material Bornyl Acetate (contains ca. 20% Isobornyl Acetate)
EINECSNumber (4)-0626&(4)-1216
TSCA Yes