missing translation for 'onlineSavingsMsg'
Learn More

(-)-Borneol 95.0+%, TCI America™

Catalog No. B101225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B101225G 25 g
B1012500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B101225G Supplier TCI America Supplier No. B101225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 464-45-9
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00003759,MFCD00066426,MFCD00066427
InChI Key DTGKSKDOIYIVQL-NQMVMOMDSA-N
Synonym --borneol, borneol, l-borneol, 1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol, linderol, 1s-endo---borneol, borneo camphor, bornyl alcohol, sumatra camphor, dl-borneol
PubChem CID 1201518
ChEBI CHEBI:15394
IUPAC Name (1S,2R,4R)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ol
SMILES CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2

Specifications

Melting Point 207°C
Color White-Yellow
Quantity 25 g
UN Number 1312
Formula Weight 154.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Borneol
EINECSNumber (4)-0607
RTECSNumber DT5095000
TSCA Yes