missing translation for 'onlineSavingsMsg'
Learn More

Boric anhydride, 99.98% trace metals basis, MilliporeSigma™ Supelco™

Catalog No. p-7187939
Change view
Click to view available options
Quantity:
100 g
500 g
Catalog No. Quantity
111012920 100 g
111012919 500 g
2 options

Catalog No. 111012920

Supplier: Merck Emd Millipore 339075100G

Only null left

Chemical Identifiers

MDL Number MFCD00011315
Synonym Boron trioxide

Specifications

Melting Point 450°C
CAS 1303-86-2
Quantity 100 g
Linear Formula B2O3
Formula Weight 69.62
Percent Purity 99.98%
Packaging Poly Bottle
Density 2.46 g/mL (at 25°C); Vapor Density: >1 (vs air)