missing translation for 'onlineSavingsMsg'
Learn More

Boric anhydride, 99.98% trace metals basis, MilliporeSigma™ Supelco™

Catalog No. 111012920
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012920 100 g
111012919 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012920 Supplier Merck Emd Millipore Supplier No. 339075100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00011315
Synonym Boron trioxide

Specifications

Melting Point 450°C
CAS 1303-86-2
Quantity 100 g
Linear Formula B2O3
Formula Weight 69.62
Percent Purity 99.98%
Packaging Poly Bottle
Density 2.46 g/mL (at 25°C); Vapor Density: >1 (vs air)