missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Borate, 0.5M buffer soln., pH 8.0

Catalog No. AAJ63742AK
Change view
Click to view available options
Quantity:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63742AK 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63742AK Supplier Thermo Scientific™ Supplier No. J63742AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 1330-43-4
Concentration 0.5M buffer soln.
pH 8.0
Quantity 250mL
MDL Number MFCD00163147
Chemical Name or Material Borate buffer soln.
Hazard Category H319-H360FD
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only