missing translation for 'onlineSavingsMsg'
Learn More

Borane-trimethylamine complex, 97%

CAS: 75-22-9 | C3H12BN | 72.95 g/mol

$171.59 - $171.59

Chemical Identifiers

CAS 75-22-9
Molecular Formula C3H12BN
Molecular Weight (g/mol) 72.95
MDL Number MFCD00012420
InChI Key WVMHLYQJPRXKLC-UHFFFAOYSA-N
IUPAC Name trimethylamine borane
SMILES B.CN(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1499414
View Documents
Thermo Scientific Chemicals
L1499414
25 g
Each for $171.59
Only null left
 
Description

Description

Reducing agent for the preparation of plate-type palladium-zinc oxide catalys, reagent for chemical vapor generation and spectrometry determination of elements using amineboranes and cyanotrihydroborate(III) reagents. Catalyst for regioselective reductive opening of benzylidene acetal glycosides and polarity reversal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reducing agent for the preparation of plate-type palladium-zinc oxide catalys, reagent for chemical vapor generation and spectrometry determination of elements using amineboranes and cyanotrihydroborate(III) reagents. Catalyst for regioselective reductive opening of benzylidene acetal glycosides and polarity reversal.

Solubility
Decomposes with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

75-22-9
72.95
WVMHLYQJPRXKLC-UHFFFAOYSA-N
B.CN(C)C
C3H12BN
MFCD00012420
trimethylamine borane

Specifications

75-22-9
0.81
64°C (147°F)
MFCD00012420
UN1325
Decomposes with water.
B.CN(C)C
72.95
97%
93°C to 95°C
172°C
C3H12BN
25 g
3905060
WVMHLYQJPRXKLC-UHFFFAOYSA-N
trimethylamine borane
72.95
Borane-trimethylamine complex
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P374

H228-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 200-850-4

RTECSNumber : PA0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only