missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Borane Tetrahydrofuran Complex Solution
Description
Borane tetrahydrofuran complex (BH3-THF) is widely used as a reducing agent in organic synthesis. It is also used as a reagent in hydroboration reactions.New, Safer, NIMBA-Stabilized BH3 THF SolutionsSure/Seal is a trademark of Sigma-Aldrich Co. LLC
Spécifications
Spécifications
| CAS | 14044-65-6 |
| Density | 0.898 g/mL at 25°C |
| Molecular Formula | C4H11BO |
| Linear Formula | BH3OC4H8 |
| MDL Number | MFCD00012429 |
| Quantity | 800 mL |
| Molecular Weight (g/mol) | 85.94 g/mol |
| Biological Activity | Respiratory System |