missing translation for 'onlineSavingsMsg'
Learn More

Borane - Tetrahydrofuran Complex (8.5% in Tetrahydrofuran, ca. 0.9mol/L), TCI America™

Catalog No. p-7137169
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
500 mL
Catalog No. Quantity
T2346100ML 100 mL
T2346500ML 500 mL
2 options

Catalog No. T2346100ML

Supplier: TCI America T2346100ML

Only null left

(stabilized with Sodium Borohydride)

Chemical Identifiers

CAS 14044-65-6
Molecular Formula C4H11BO
Molecular Weight (g/mol) 85.94
MDL Number MFCD00012429
InChI Key RMCYTHFAWCWRFA-UHFFFAOYSA-N
Synonym borane-tetrahydrofuran complex, tetrahydrofuran borane, bh3.thf, borane tetrahydrofuran complex solution, borane-d3-thf complex solution, borane-tetrahydrofuran, unii-5ear4err1l, oxolane borane, boron; oxolane, borane thf
PubChem CID 11062302
IUPAC Name oxolane borane
SMILES B.C1CCOC1

Specifications

Melting Point -17°C
Color Yellow
Quantity 100 mL
UN Number 3399
Formula Weight 85.94
Physical Form Liquid
Chemical Name or Material Borane - Tetrahydrofuran Complex (8.5% in Tetrahydrofuran, ca. 0.9mol/L) (stabilized with Sodium Bor
TSCA Yes
Recommended Storage Refrigerator