missing translation for 'onlineSavingsMsg'
Learn More

Borane - Tetrahydrofuran Complex (8.5% in Tetrahydrofuran, ca. 0.9mol/L), TCI America™

Catalog No. T2346100ML
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2346100ML 100 mL
T2346500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2346100ML Supplier TCI America Supplier No. T2346100ML

(stabilized with Sodium Borohydride)

Chemical Identifiers

CAS 14044-65-6
Molecular Formula C4H11BO
Molecular Weight (g/mol) 85.94
MDL Number MFCD00012429
InChI Key RMCYTHFAWCWRFA-UHFFFAOYSA-N
Synonym borane-tetrahydrofuran complex, tetrahydrofuran borane, bh3.thf, borane tetrahydrofuran complex solution, borane-d3-thf complex solution, borane-tetrahydrofuran, unii-5ear4err1l, oxolane borane, boron; oxolane, borane thf
PubChem CID 11062302
IUPAC Name oxolane borane
SMILES B.C1CCOC1

Specifications

Melting Point -17°C
Color Yellow
Quantity 100 mL
UN Number 3399
Formula Weight 85.94
Physical Form Liquid
Chemical Name or Material Borane - Tetrahydrofuran Complex (8.5% in Tetrahydrofuran, ca. 0.9mol/L) (stabilized with Sodium Bor
TSCA Yes
Recommended Storage Refrigerator