missing translation for 'onlineSavingsMsg'
Learn More

Borane-tert-butylamine complex, 97%

Catalog No. AAL1317709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1317709 10 g
AAL1317718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1317709 Supplier Thermo Scientific Chemicals Supplier No. L1317709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7337-45-3 | C4H14BN | 86.97 g/mol

Borane-tert-butylamine complex is used in stereoselective reduction of a steroidal ketone and for a review of amine boranes as selective reducing and hydroborating agents. It is also used as strong co-reducing agent in the syntheses of copper (I) oxide nanoparticles, diphosphine-protected gold nanocluster and 4-acetoxycinnamyl alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Borane-tert-butylamine complex is used in stereoselective reduction of a steroidal ketone and for a review of amine boranes as selective reducing and hydroborating agents. It is also used as strong co-reducing agent in the syntheses of copper (I) oxide nanoparticles, diphosphine-protected gold nanocluster and 4-acetoxycinnamyl alcohols.

Solubility
Soluble in water (27 g/L at 20°C) and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and acids.

Chemical Identifiers

CAS 7337-45-3
Molecular Formula C4H14BN
Molecular Weight (g/mol) 86.97
MDL Number MFCD00075635
InChI Key GKFJEDWZQZKYHV-UHFFFAOYSA-N
Synonym tert-butylamine borane, borane-tert-butylamine complex, borane tert-butylamine complex, borane-tert-butylaminecomplex, c4h14bn, borane t-butylamine, t-butylamine.borane, tert-butylamineborane, borane tert-butylamine, tert-butylamine-borane
PubChem CID 6364547
IUPAC Name boron;2-methylpropan-2-amine
SMILES B.CC(C)(C)N

Specifications

Melting Point 96°C to 98°C
Density 0.87
Quantity 10 g
UN Number UN2811
Beilstein 4134905
Solubility Information Soluble in water (27g/L at 20°C) and methanol.
Formula Weight 86.98
Percent Purity 97%
Chemical Name or Material Borane-tert-butylamine complex
Hazard Category H228-H301+H311-H314-H332-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 230-851-5
RTECSNumber EO3900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only