missing translation for 'onlineSavingsMsg'
Learn More

Borane-N,N-diethylaniline complex, 97%

Catalog No. p-3735678
Change view
Click to view available options
Quantity:
100 g
Packaging:
AcroSeal™ Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC176771000 100 g AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC176771000 Supplier Thermo Scientific Chemicals Supplier No. 176771000

CAS: 13289-97-9 | C10H15BN | 160.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 13289-97-9 , 91-66-7
Molecular Formula C10H15BN
Molecular Weight (g/mol) 160.05
MDL Number MFCD00013187
InChI Key KHYAFFAGZNCWPT-UHFFFAOYSA-N
Synonym n,n-diethylaniline borane, borane-n,n-diethylaniline complex, borane-n,n-diethylaniline, n,n-diethylanilineborane, diethylaniline borane, n,n-diethylaniline trihydroboron, n.n diethylanilineborane, boron; n,n-diethylaniline, boron, n,n-diethylbenzenamine trihydro-, t-4, borane n,n-diethylaniline complex
PubChem CID 6335292
IUPAC Name boron;N,N-diethylaniline
SMILES [B].CCN(CC)C1=CC=CC=C1

Specifications

Melting Point -30°C to 27°C
Color Colorless to Amber
Density 0.9200g/mL
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging AcroSeal™ Glass bottle
Refractive Index 1.5245 to 1.5265
Linear Formula C6H5N(C2H5)2·BH3
Quantity 100 g
Specific Gravity 0.92
Solubility Information Solubility in water: may decompose
Formula Weight 163.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Borane-N, N-diethylaniline complex
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
In contact with wat
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 236-305-2

RUO – Research Use Only