missing translation for 'onlineSavingsMsg'
Learn More

Borane-dimethylamine complex, 97%

Catalog No. p-7080498
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8914914 25 g
AA8914922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8914914 Supplier Thermo Scientific Chemicals Supplier No. 08914914
Only null left

CAS: 74-94-2 | C2H10BN | 58.92 g/mol

Borane-dimethylamine complex is used in a nickel-catalyzed reduction of electron-deficient aryl tosylates. It also serves as a reducing agent in electrolytic deposition of metals, alloys, semiconductors and insulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74-94-2
Molecular Formula C2H10BN
Molecular Weight (g/mol) 58.92
MDL Number MFCD00051068
InChI Key FTDUHBOCJSQEKS-UHFFFAOYSA-N
Synonym Dimethylamine borane
IUPAC Name dimethylamine borane
SMILES B.CNC

Specifications

Melting Point 36°C
Color White
Density 0.69 g/mL
Boiling Point 59°C to 65°C (1 to 2 mmHg)
Quantity 25 g
UN Number UN2925
Sensitivity Air and moisture sensitive
Solubility Information Soluble in methanol,water,ethanol,2-propanol and terahydrofuran.
Formula Weight 58.92
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Borane-dimethylamine complex
Show More Show Less
Hazard Category H228-H301+H311-H315-H319-H332-H335
Hazard Statement GHS H Statement
H301-H310-H330-H314-H318-H228
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 200-823-7
RTECSNumber IP9450000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only