missing translation for 'onlineSavingsMsg'
Learn More

Borane-dimethyl sulfide complex, 94%

Catalog No. AAL07705AE
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL07705AE 100 mL
AAL07705AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL07705AE Supplier Thermo Scientific Chemicals Supplier No. L07705AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13292-87-0 | C2H9BS | 75.96 g/mol

Borane- dimethyl sulfide is used for hydroboration and reduction reactions. It is used in the reduction of aldehydes, ketones, epoxides and carboxylic acids to give corresponding alcohols. It is also involved in the Corey-Itsuno reduction. It acts as an intermediate in the preparation of highly pure semiconductor. It is also employed as a rocket propellant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Borane- dimethyl sulfide is used for hydroboration and reduction reactions. It is used in the reduction of aldehydes, ketones, epoxides and carboxylic acids to give corresponding alcohols. It is also involved in the Corey-Itsuno reduction. It acts as an intermediate in the preparation of highly pure semiconductor. It is also employed as a rocket propellant.

Solubility
Miscible with ethyl ether, dichloromethane, benzene, xylene, hexane, diglyme, dimethyl ether, ethyl acetate, toluene, methylene chloride and other aprotic solvents.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Store in a cool place. Incompatible with acids, water, alcohols, bromine, acid anhydrides, acid chlorides and oxidizing agents.

Chemical Identifiers

CAS 13292-87-0
Molecular Formula C2H9BS
Molecular Weight (g/mol) 75.96
MDL Number MFCD00013189
InChI Key RMHDLBZYPISZOI-UHFFFAOYSA-N
Synonym dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
PubChem CID 9833925
IUPAC Name boron;methylsulfanylmethane
SMILES B.CSC

Specifications

Melting Point -38°C
Density 0.801
Boiling Point ∼44°C (decomposition)
Flash Point −1°C (30°F)
Refractive Index 1.457
Linear Formula (CH3)2S·BH3
Quantity 100 mL
UN Number UN2924
Beilstein 3663489
Sensitivity Air and moisture sensitive
Solubility Information Miscible with ethyl ether,dichloromethane,benzene,xylene,hexane,diglyme,dimethyl ether,ethyl acetate,toluene,methylene chloride and other aprotic solvents.
Formula Weight 75.97
Percent Purity 94%
Chemical Name or Material Borane-dimethyl sulfide complex
Show More Show Less
Hazard Category H225-H260-H301-H315-H318
Hazard Statement GHS H Statement
H225-H260-H300-H314-H318-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement P210-P223-P231+P232-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P310-P303+P361+P353-P305+P351+P338-P310-P330-P332+P313-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 236-313-6
RTECSNumber PV5080000
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only