missing translation for 'onlineSavingsMsg'
Learn More

Borane - 2-Methylpyridine Complex 85.0+%, TCI America™

Catalog No. p-7119896
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30185G 5 g
B301825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30185G Supplier TCI America Supplier No. B30185G
Only null left

Chemical Identifiers

CAS 3999-38-0
Molecular Formula C6H10BN
Molecular Weight (g/mol) 106.96
MDL Number MFCD07784361
InChI Key ZMASBEFBIXMNCP-UHFFFAOYSA-N
Synonym Borane - 2-Picoline Complex, 2-Picoline Borane, 2-Methylpyridine Borane
IUPAC Name 2-methylpyridine borane
SMILES B.CC1=CC=CC=N1

Specifications

Melting Point 48°C
Color White-Yellow
Quantity 5 g
UN Number 2813
Formula Weight 106.96
Percent Purity ≥85.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Borane - 2-Methylpyridine Complex