missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bopindolol malonate, 97+%

Catalog No. aaj67119mc
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67119MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67119MC Supplier Thermo Scientific Chemicals Supplier No. J67119MC

CAS: 62658-64-4 | C23H28N2O3.C3H4O4, C26H32N2O7 | 484.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62658-64-4
Molecular Formula C26H32N2O7
Molecular Weight (g/mol) 484.55
InChI Key LDOONRJGLKHGJI-UHFFFAOYNA-N
IUPAC Name 1-(tert-butylamino)-3-[(2-methyl-1H-indol-4-yl)oxy]propan-2-yl benzoate; propanedioic acid
SMILES OC(=O)CC(O)=O.CC1=CC2=C(OCC(CNC(C)(C)C)OC(=O)C3=CC=CC=C3)C=CC=C2N1

Specifications

Quantity 100 mg
Description White
Formula Weight 484.54
Product Type Bopindolol malonate, 99%
Form Solid
Assay 99%
Recommended Storage Ambient temperatures

RUO – Research Use Only