missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bombesin

Catalog No. p-5384573
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63514MCR 1 mg
AAJ63514LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63514MCR Supplier Thermo Scientific Chemicals Supplier No. J63514MCR
Only null left

For cellular and molecular biology applications

Gut tetradecapeptide with the ability to stimulate release of various hormones

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Purification Method HPLC
Content And Storage 2°C to 8°C
Format Powder
For Use With (Application) Gut tetradecapeptide with the ability to stimulate release of various hormones
Molecular Weight (g/mol) 1619.90
Product Type Bombesin
Quantity 1 mg
Regulatory Status RUO - research use only
Sequence Glp-Gln-Arg-Leu-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2

RUO – Research Use Only