missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bolt™ Welcome Pack with iBlot™ 3 System
GREENER_CHOICE

Catalog No. nw0412aib3
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NW0412AIB3 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NW0412AIB3 Supplier Invitrogen™ Supplier No. NW0412AIB3

The Bolt Welcome Pack with iBlot 3 System offers a complete protein separation-to-western blot solution by combining a Bolt Welcome Pack A with the iBlot 3 Western Blot Transfer Device and iBlot 3 Transfer Stacks, Midi, NC (nitrocellulose) in lieu of one box of Bolt gels.

The Bolt Welcome Pack with iBlot 3 System offers a complete protein separation-to-western blot solution by combining a Bolt Welcome Pack A with the iBlot 3 Western Blot Transfer Device and iBlot 3 Transfer Stacks, Midi, NC (nitrocellulose) in lieu of one box of Bolt gels.

The Bolt Welcome Pack A consists of a Mini Gel Tank plus all of the necessary Bolt gels and buffers you need to get started running gels.

Contents of the Bolt Welcome Pack with iBlot 3 System:

  • iBlot 3 Western Blot Transfer Device
  • iBlot 3 Transfer Stacks, Midi, NC (1 box)
  • Mini Gel Tank
  • Bolt 4-12% Bis-Tris Plus gels, 10-well (1 box)
  • Bolt MES Running Buffer (20X, 500 mL)
  • MES and MOPS Running Buffer, Powder (NP000105, NP000205)
  • Bolt LDS Sample Buffer (4X, 10 mL)
  • Bolt Sample Reducing Agent (10X, 10 mL)
  • PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (2 x 250 μL)

About the Mini Gel Tank

The Mini Gel Tank is compatible with all Novex, NuPAGE, and Bolt Bis-Tris Plus mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt Bis-Tris Plus gels to be run in approximately 20 minutes with MES running buffer. Run times may vary depending on buffer conditions and power supplies being used.

About Bolt Bis-Tris Plus Gels

Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal broad range separation of proteins under denaturing conditions. Similar to NuPAGE Bis-Tris gels, Bolt Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt Bis-Tris Plus gels have the unique WedgeWell design that allows for easier sample loading and up to 2X more sample volume than other standard precast gels. Bolt Bis-Tris Plus gels come in four percentages and multiple well formats. Each box contains 10 gels.

About the iBlot 3 Western Blot Transfer Device

The iBlot 3 Western Blot Transfer Device is a dry transfer device that performs western blotting transfer efficiently and reliably in as few as three minutes and without the need for liquid buffers. The iBlot 3 system is compatible with both nitrocellulose and polyvinylidene difluoride (PVDF) membranes and offers equivalent or better performance compared with traditional wet-transfer methods in a fraction of the time. The iBlot 3 Western Blot Transfer Device is an integral part of the iBlot dry blotting system, which consists of the transfer device and consumable transfer stacks that contain the required buffers and transfer membrane (nitrocellulose or PVDF).

Specifications

Content And Storage • iBlot 3 Gel Transfer Device
• Western Blot Roller
• iBlot 3 Transfer Stacks, midi, nitrocellulose
• Mini Gel Tank
• Bolt 4-12% Bis-Tris Plus gels, 10-well (1 box)
• Bolt MES Running Buffer (20X, 500 mL)
• Bolt LDS Sample Buffer (4X, 10 mL)
• Bolt Sample Reducing Agent (10X, 10 mL)
• PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (2 x 250 μL)

For Use With (Application) Western Blot
Gel Size Mini
Membrane Compatibility Nitrocellulose, PVDF
Product Line Bolt, iBlot 3
Gel Percentage 4 to 12%
Gel Type Bis-Tris Plus
No. of Wells 10-well
Quantity 1 each
Separation Range 3.5 to 260 kd
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Thickness (Metric) 1.0 mm
Show More Show Less
Do you offer additional years of service for the iBlot 3 Western Transfer Device?

No, we do not provide service contracts or extended warranties for these devices. The warranty is limited to 1 year from the date of shipment.

For Research Use Only. Not for use in diagnostic procedures.