missing translation for 'onlineSavingsMsg'
Learn More

Boekel Scientific™ TropiCooler™ Benchtop Hot/Cold Block Incubator

Catalog No. 054507
Change view
Click to view available options
Temperature Range (Metric):
-19°C to +69°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
054507 -19°C to +69°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 054507 Supplier Boekel Scientific™ Supplier No. 260014

Includes

  • Two machined aluminum blocks, holding 14 microcentrifuge tubes (1.5mL size) each (additional block configurations available)
  • Smoked acrylic plastic lid

Store frozen restriction enzymes and perform ligations and DNA hybridizations from the bench

  • Temperature range: -19° to +69°C (-2° to +156°F)
  • Easy temperature selection (settings in 1° increments) and rapid equilibration time Digital microprocessor controls Peltier thermoelectric heating and cooling device for efficient, low-maintenance operation
  • Dry heat and cold make unit an attractive alternative to ice buckets and water baths

Order Info

Dimensions, 11 3/4L x 9 3/4W x 7 1/4 in.H (29.8 x 24.8 x 18.4cm); Weight, 13 lb. (6kg); For 115V 60Hz, 0.8A, 100w

Service Plan

Service agreement available.

Specifications

Certifications/Compliance CE
For Use With (Equipment) Storing frozen restriction Enzymes or DNA libraries
No. of Places 2
Temperature Range (Metric) -19°C to +69°C
Display LCD
Includes Two machined Aluminum blocks and smoked Acrylic Plastic flat lid
Temperature Accuracy ±1°C
Warranty 2 Years
Dimensions (L x W x H) 11.75 x 9.75 x 7.25 in. (28.6 x 24.8 x 18.4 cm)
Length (English) 11.75 in.
Length (English) Sink 3.5 in.
Length (Metric) 28.6 cm
Length (Metric) Sink 8.7 cm
Width (English) 9.75 in.
Width (Metric) 24.8 cm
Height (English) 7.25 in.
Height (Metric) 18.4 cm
Shipping Weight (English) 17 lb.
Shipping Weight (Metric) 7.7 kg
Electrical Requirements 115 V, 50/60 Hz
Voltage 115 V
Wattage 100 w
Amperage 0.8 A
Frequency 50/60 Hz
Show More Show Less