missing translation for 'onlineSavingsMsg'
Learn More

Boekel Scientific™ Jitterbug™ Microplate Incubator Shaker
GREENER_CHOICE

Catalog No. 1170113
Change view
Click to view available options
Speed Range:
575 to 1500 rpm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Speed Range
1170113 575 to 1500 rpm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1170113 Supplier Boekel Scientific™ Supplier No. 130000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Jitterbug™ Microplate Incubator Shaker accepts two standard microplates or strip trays.

  • Ideal for a variety of immunoassay and research applications.
  • Microprocessor allows independent control of temperature, shaking speed, and mixing duration.
  • Warmed inner aluminum bed with air circulation provides temperature uniformity better than ±0.4°C (with cover).
  • Inner chamber cover insulates plates, reduces biohazard risk; outer tinted plexiglass cover adds insulation and protection from light.
  • Automatic circulating fan prevents condensation.
  • Rotating counterweight mixer offers continuous timed operation (575 to 1500 rpm in 8 increments) or pulse operation (1500 rpm for 3 sec.).
  • Timer has 1-second resolution and range of 1 second to 99 minutes, 59 seconds.
  • Temperature range is ambient to 40°C with resolution of 0.1°C; warms up in 15 minutes.
  • ABS plastic enclosure with a metal base.
  • Orbit Variable 1 to 2 mm.

Order Info

Dimensions (L x W x H): 10 x 10 x 4 in. (25.4 x 25.4 x 10.2 cm). Net weight: 7 lb. (3.2 kg). For 115 V, supplied with 120 VAC input, 12 VAC 2 A output adapter.

Specifications

Orbit 1 mm
Rotating Angle 360°
Load Bearing Capacity (Metric) 0.3 kg
Speed Range 575 to 1500 rpm
Electrical Requirements 115 V, 50/60 Hz
Voltage 115 VAC
Temperature Range (Metric) 40°C
Wattage 33 W
Amperage 1 A
Load Bearing Capacity (English) 0.66 lb.
Certifications/Compliance CE, CSA, UL
Temperature Range (English) 104°F
Frequency 50/60 Hz
Depth (English) 10 in.
Depth (Metric) 25.4 cm
Height (English) 4 in.
Height (Metric) 10.2 cm
Length (English) 10 in.
Length (Metric) 25.4 cm
Width (English) 10 in.
Width (Metric) 25.4 cm
Shipping Weight (English) 11 lb.
Shipping Weight (Metric) 4.99 kg
Model Jitterbug
For Use With (Application) Immunoassay PCR and other research procedures requiring microplates
Warranty and Service Offering 2 Year
Show More Show Less