Learn More
Boekel Scientific™ InSlide Out™ Hybridization Apparatus Polycarbonate Slide Holders
Catalog No. 0545019
Change view
Click to view available options
Material:
Polycarbonate
Catalog No. Material
0545019 Polycarbonate
1 options
Catalog No. 0545019 Supplier Boekel Scientific™ Supplier No. C2403765/DUP
Only null left
Holds up to eighteen 1 x 3 in. slides only
  • Holds up to eighteen 1 × 3 in. slides only
  • Slides are placed in slots for ease in carrying
Specifications
For Use With (Equipment) InSlide out hybridization apparatus
Holds Eighteen 1 x 3 in. slide
Material Polycarbonate
Type Slide Holder
For Use With (Application) InSlide out hybridization apparatus