missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ BODIPY™ TMR C5-Maleimide

Catalog No. b30466
Encompass
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
Catalog No. Quantity
B30466 1 mg
1 options

Catalog No. B30466

Supplier: Molecular Probes™ B30466

Only null left

BODIPY™ TMR C5-maleimide

The thiol-reactive BODIPY™ TMR maleimide produces electronically neutral dye conjugates that are spectrally similar to the positively charged tetramethylrhodamine dye. This dye's lack of ionic charge results in minimal effects on the isoelectric points of standard proteins conjugated with this fluorophore. The small size and relatively long excited-state lifetime of BODIPY™ TMR dye has proven useful for studying ligand-receptor interaction by fluorescence polarization.

Specifications

Quantity 1 mg
Product Type Maleimide
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Maleimide
Label or Dye BODIPY™ TMR
Label Type BODIPY Dyes
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.