missing translation for 'onlineSavingsMsg'
Learn More

BOC-N-Methyl-L-alanine, 99%

Catalog No. AC442930010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442930010 1 g
AC442930050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442930010 Supplier Thermo Scientific Chemicals Supplier No. 442930010

CAS: 16948-16-6 | C9H17NO4 | 203.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16948-16-6
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00037242
InChI Key VLHQXRIIQSTJCQ-LURJTMIESA-N
Synonym boc-n-me-ala-oh, boc-n-methyl-l-alanine, n-boc-n-methyl-l-alanine, s-2-tert-butoxycarbonyl methyl amino propanoic acid, n-tert-butoxycarbonyl-n-methyl-l-alanine, boc-n-a-methyl-l-alanine, s-2-tert-butoxycarbonyl-methyl-amino-propionic acid, l-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2s-2-tert-butoxycarbonyl methyl amino propanoic acid, 2s-2-tert-butoxy carbonyl methyl amino propanoic acid
PubChem CID 7009590
IUPAC Name (2S)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
SMILES C[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 88.0°C to 92.0°C
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 1 g
Specific Rotation − 29.80
Formula Weight 203.24
Percent Purity 99%
Chemical Name or Material BOC-N-Methyl-L-alanine
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only