missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Valine, 99+%

CAS: 13734-41-3 | C10H19NO4 | 217.27 g/mol

$368.49 - $368.49

Chemical Identifiers

CAS 13734-41-3
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00065605
InChI Key SZXBQTSZISFIAO-ZETCQYMHSA-N
Synonym boc-val-oh, boc-l-valine, n-tert-butoxycarbonyl-l-valine, tert-butoxycarbonylvaline, boc-l-val-oh, s-2-boc-amino-3-methylbutyric acid, s-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, n-boc-l-valine, n-tert-butyloxycarbonyl-l-valine
PubChem CID 83693
SMILES CC(C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC275711000
SDS View Documents
Thermo Scientific Chemicals
275711000
100 g Glass Bottle
Each for $368.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13734-41-3
217.27
SZXBQTSZISFIAO-ZETCQYMHSA-N
83693
C10H19NO4
MFCD00065605
boc-val-oh, boc-l-valine, n-tert-butoxycarbonyl-l-valine, tert-butoxycarbonylvaline, boc-l-val-oh, s-2-boc-amino-3-methylbutyric acid, s-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, n-boc-l-valine, n-tert-butyloxycarbonyl-l-valine
CC(C)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

76°C to 80°C
White
99+%
Glass Bottle
MFCD00065605
boc-val-oh, boc-l-valine, n-tert-butoxycarbonyl-l-valine, tert-butoxycarbonylvaline, boc-l-val-oh, s-2-boc-amino-3-methylbutyric acid, s-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, n-boc-l-valine, n-tert-butyloxycarbonyl-l-valine
CC(C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
- 6.30 (20.00°C c=1,CH3COOH)
100 g
217.27
Fine Crystalline Powder
13734-41-3
Authentic
C10H19NO4
(CH3)2CHCHCOOHNHCOOC(CH3)3
-5.3° to -7.3° (20°C, 589nm) (c=1, CH3COOH)
SZXBQTSZISFIAO-ZETCQYMHSA-N
(2S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
217.27
83693
≥99.0%
BOC-L-Valine, 99+%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 237-307-6

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only