missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Valine, 99+%

Catalog No. AC275710050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC275710050 5 g Glass Bottle
AC275710250 25 g Glass Bottle
AC275711000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC275710050 Supplier Thermo Scientific Chemicals Supplier No. 275710050

CAS: 13734-41-3 | C10H19NO4 | 217.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13734-41-3
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00065605
InChI Key SZXBQTSZISFIAO-ZETCQYMHSA-N
Synonym boc-val-oh, boc-l-valine, n-tert-butoxycarbonyl-l-valine, tert-butoxycarbonylvaline, boc-l-val-oh, s-2-boc-amino-3-methylbutyric acid, s-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, n-boc-l-valine, n-tert-butyloxycarbonyl-l-valine
PubChem CID 83693
SMILES CC(C)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 76°C to 80°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula (CH3)2CHCHCOOHNHCOOC(CH3)3
Specific Rotation -5.3° to -7.3° (20°C, 589nm) (c=1, CH3COOH)
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoic acid
Quantity 5 g
Formula Weight 217.27
Percent Purity ≥99.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material BOC-L-Valine, 99+%
Show More Show Less
EINECSNumber 237-307-6
TSCA TSCA

RUO – Research Use Only