missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Tyrosine, 99+%

Catalog No. AC275700050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC275700050 5 g Glass Bottle
AC275700250 25 g Glass Bottle
AC275701000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC275700050 Supplier Thermo Scientific Chemicals Supplier No. 275700050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3978-80-1 | C14H19NO5 | 281.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3978-80-1
Molecular Formula C14H19NO5
Molecular Weight (g/mol) 281.31
MDL Number MFCD00037179
InChI Key CNBUSIJNWNXLQQ-LDGXTIHJNA-N
Synonym boc-tyr-oh, boc-l-tyrosine, n-boc-l-tyrosine, n-tert-butoxycarbonyl-l-tyrosine, n-boc-tyrosine, n-t-boc-l-tyrosine, l-tyrosine, n-1,1-dimethylethoxy carbonyl, chembl65386, n-t-butoxycarbonyl-l-tyrosine, n-tert-butoxycarbonyl tyrosine
PubChem CID 117439
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Melting Point 135.0°C to 140.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula HOC6H4CH2CH(COOH)NHCO2C(CH3)3
Specific Rotation + 2.50
Water 1% Max. (K.F.)
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
Quantity 5 g
Formula Weight 281.3
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material BOC-L-Tyrosine
Show More Show Less

RUO – Research Use Only