missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-alpha-phenylglycine, 99%

Catalog No. AC398160010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398160010 1 g Glass Bottle
AC398160050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC398160010 Supplier Thermo Scientific Chemicals Supplier No. 398160010
Only null left

CAS: 2900-27-8 | C13H17NO4 | 251.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2900-27-8
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00065588
InChI Key HOBFSNNENNQQIU-JTQLQIEISA-N
Synonym boc-phg-oh, boc-l-phenylglycine, s-2-tert-butoxycarbonyl amino-2-phenylacetic acid, boc-l-alpha-phenylglycine, boc-l-phg-oh, boc-l-a-phenylglycine, n-boc-l-alpha-phenylglycine, n-boc-l-2-phenylglycine, s-boc-l-phenylglycine, n-boc-l-phenylglycine
PubChem CID 11010409
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-phenylacetic acid
SMILES CC(C)(C)OC(=O)NC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 88°C to 91°C
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 251.28
Percent Purity 99%
Chemical Name or Material BOC-L-α-phenylglycine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only