missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Phenylalanine, 99+%

CAS: 13734-34-4 | C14H19NO4 | 265.31 g/mol

Supplier:  Thermo Scientific Chemicals 275640250

Quantity 25 g
Packaging Glass Bottle

 View more versions of this product

Catalog No. AC275640250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13734-34-4
265.31
ZYJPUMXJBDHSIF-NSHDSACASA-N
77857
C14H19NO4
MFCD00002663
boc-phe-oh, boc-l-phenylalanine, boc-phenylalanine, n-tert-butoxycarbonyl-l-phenylalanine, n-boc-l-phenylalanine, n-t-boc-l-phenylalanine, n-alpha-t-boc-l-phenylalanine, n-t-butyloxycarbonyl-l-phenylalanine, l-phenylalanine, n-1,1-dimethylethoxy carbonyl, boc-phe
CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

BOC-L-Phenylalanine
13734-34-4
Authentic
Glass Bottle
C6H5CH2CH(COOH)NHCOOC(CH3)3
+ 25.00
boc-phe-oh, boc-l-phenylalanine, boc-phenylalanine, n-tert-butoxycarbonyl-l-phenylalanine, n-boc-l-phenylalanine, n-t-boc-l-phenylalanine, n-alpha-t-boc-l-phenylalanine, n-t-butyloxycarbonyl-l-phenylalanine, l-phenylalanine, n-1,1-dimethylethoxy carbonyl, boc-phe
0.5% max. (K.F.)
CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
25 g
77857
≥99%
85.0°C to 88.0°C
White
99+%
C14H19NO4
MFCD00002663
14, 1595
Solubility in water: insoluble. Other solubilities: soluble in ethanol and methylene chloride, insoluble in aliphatic hydrocarbons
ZYJPUMXJBDHSIF-NSHDSACASA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
265.31
265.3
Fine Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions