missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Methionine, 98%

CAS: 2488-15-5 | C10H19NO4S | 249.33 g/mol

$71.81 - $179.49

Chemical Identifiers

CAS 2488-15-5
Molecular Formula C10H19NO4S
Molecular Weight (g/mol) 249.33
MDL Number MFCD00065586
InChI Key IMUSLIHRIYOHEV-ZETCQYMHSA-N
Synonym boc-l-methionine, boc-met-oh, n-boc-l-methionine, n-tert-butoxycarbonyl-l-methionine, tert-butoxycarbonyl-l-methionine, s-2-tert-butoxycarbonyl amino-4-methylthio butanoic acid, n-alpha-t-butyloxycarbonyl-l-methionine, methionine, n-1,1-dimethylethoxy carbonyl, l-methionine, n-1,1-dimethylethoxy carbonyl
PubChem CID 89857
SMILES CSCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC275630050
SDS View Documents
Thermo Scientific Chemicals
275630050
5 g Glass Bottle
Each for $71.81
Only null left
 
AC275630250
SDS View Documents
Thermo Scientific Chemicals
275630250
25 g Glass Bottle
Each for $179.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2488-15-5
249.33
IMUSLIHRIYOHEV-ZETCQYMHSA-N
89857
C10H19NO4S
MFCD00065586
boc-l-methionine, boc-met-oh, n-boc-l-methionine, n-tert-butoxycarbonyl-l-methionine, tert-butoxycarbonyl-l-methionine, s-2-tert-butoxycarbonyl amino-4-methylthio butanoic acid, n-alpha-t-butyloxycarbonyl-l-methionine, methionine, n-1,1-dimethylethoxy carbonyl, l-methionine, n-1,1-dimethylethoxy carbonyl
CSCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

47.0°C to 53.0°C
White
99+%
Glass Bottle
MFCD00065586
boc-l-methionine, boc-met-oh, n-boc-l-methionine, n-tert-butoxycarbonyl-l-methionine, tert-butoxycarbonyl-l-methionine, s-2-tert-butoxycarbonyl amino-4-methylthio butanoic acid, n-alpha-t-butyloxycarbonyl-l-methionine, methionine, n-1,1-dimethylethoxy carbonyl, l-methionine, n-1,1-dimethylethoxy carbonyl
IMUSLIHRIYOHEV-ZETCQYMHSA-N
(2S)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
249.33
249.33
Powder
2488-15-5
Authentic
C10H19NO4S
CH3SCH2CH2CH(COOH)NHCOOC(CH3)3
− 23.00
0.5% max. (K.F.)
CSCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
5 g
89857
≥99%
BOC-L-Methionine, 99+%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-639-3

SDS
Documents

Documents

RUO – Research Use Only