missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-alpha-cyclohexylglycine, 98%

Catalog No. AC399350010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399350010 1 g Glass Bottle
AC399350050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399350010 Supplier Thermo Scientific Chemicals Supplier No. 399350010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109183-71-3 | C13H23NO4 | 257.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109183-71-3
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.32
MDL Number MFCD00133638
InChI Key QSUXZIPXYDQFCX-JTQLQIEISA-N
Synonym boc-l-cyclohexylglycine, boc-chg-oh, boc-l-alpha-cyclohexylglycine, n-boc-2-cyclohexyl-l-glycine, s-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-cyclohexyl-glycine, 2s-tert-butoxycarbonyl amino cyclohexyl acetic acid, s-tert-butoxycarbonylamino-cyclohexyl-acetic acid, s-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2s-tert-butoxycarbonyl amino cyclohexyl ethanoic acid
PubChem CID 7004938
IUPAC Name (2S)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
SMILES CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O

Specifications

Melting Point 72°C to 76°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation +8° to +11° (20°C, 589nm) (c=1, CH3OH)
Formula Weight 257.32
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material BOC-L-α-Cyclohexylglycine
Show More Show Less

RUO – Research Use Only