missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Aspartic acid 4-benzylester, 99+%, Thermo Scientific™

$104.59

Chemical Identifiers

CAS 7536-58-5
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.345
MDL Number MFCD00065564
InChI Key SOHLZANWVLCPHK-LBPRGKRZSA-N
Synonym boc-asp obzl-oh, boc-l-aspartic acid 4-benzyl ester, s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp obzl-oh, boc-l-aspartic acid-4-benzyl ester, 4-benzyl n-boc-l-aspartate, boc-l-aspartic acid 4-benzylester, 2s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-aspartic acid beta-benzyl ester, n-tert-butoxycarbonyl-l-aspartic acid 4-benzyl ester
PubChem CID 1581888
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC275520050
View Documents Promotion Details
N/A 5 g Glass Bottle
Each for $104.59
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7536-58-5
323.345
SOHLZANWVLCPHK-LBPRGKRZSA-N
1581888
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
C16H21NO6
MFCD00065564
boc-asp obzl-oh, boc-l-aspartic acid 4-benzyl ester, s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp obzl-oh, boc-l-aspartic acid-4-benzyl ester, 4-benzyl n-boc-l-aspartate, boc-l-aspartic acid 4-benzylester, 2s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-aspartic acid beta-benzyl ester, n-tert-butoxycarbonyl-l-aspartic acid 4-benzyl ester
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid

Specifications

98°C to 102°C
White
99+%
C6H5CH2OCOCH2CH(COOH)nHCOOC(CH3)3
-18.5° to -21° (20°C, 589nm) (c=2, DMF)
SOHLZANWVLCPHK-LBPRGKRZSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
1581888
≥99.0%
BOC-L-Aspartic acid 4-benzylester
7536-58-5
Authentic
C16H21NO6
MFCD00065564
boc-asp obzl-oh, boc-l-aspartic acid 4-benzyl ester, s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp obzl-oh, boc-l-aspartic acid-4-benzyl ester, 4-benzyl n-boc-l-aspartate, boc-l-aspartic acid 4-benzylester, 2s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-aspartic acid beta-benzyl ester, n-tert-butoxycarbonyl-l-aspartic acid 4-benzyl ester
CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O
323.345
323.34
Powder
Videos
SDS
Documents

Documents

RUO – Research Use Only