missing translation for 'onlineSavingsMsg'
Learn More

BOC-DL-Valine, 98%

Catalog No. AC425780100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425780100 10 g Glass bottle
AC425780500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425780100 Supplier Thermo Scientific Chemicals Supplier No. 425780100
Only null left

CAS: 54895-12-4 | C10H19NO4 | 217.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54895-12-4
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00069973
InChI Key SZXBQTSZISFIAO-UHFFFAOYSA-N
Synonym boc-dl-valine, boc-dl-val-oh, n-boc-dl-valine, 2-tert-butoxycarbonyl amino-3-methylbutanoic acid, dl-n-tert-butoxycarbonyl valine, n-tert-butoxycarbonyl valine, 2-tert-butoxy carbonyl amino-3-methylbutanoic acid, n-tert-butoxycarbonyl-dl-valine, n-alpha-t-boc-l-valine
PubChem CID 270655
IUPAC Name 3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)C(NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 112°C to 114°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH(COOH)NHCOOC(CH3)3
Quantity 10 g
Formula Weight 217.26
Percent Purity 98%
Chemical Name or Material BOC-DL-Valine

RUO – Research Use Only