missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOC-D-α-cyclohexylglycine, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 70491-05-3 |
|---|---|
| Molecular Formula | C13H23NO4 |
| Molecular Weight (g/mol) | 257.33 |
| MDL Number | MFCD00133629 |
| InChI Key | QSUXZIPXYDQFCX-SNVBAGLBSA-N |
| Synonym | boc-d-chg-oh, boc-d-cyclohexylglycine, r-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-alpha-cyclohexyl-d-glycine, n-boc-2-cyclohexyl-d-glycine, 2r-n-tert-butoxycarbonyl-2-cyclohexylglycine, r-tert-butoxycarbonylamino-cyclohexyl-acetic acid, r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-2-tert-butoxycarbonylamino-2-cyclohexyl-acetic acid |
| PubChem CID | 7014890 |
| IUPAC Name | (2R)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid |
| SMILES | CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC399360010
|
Thermo Scientific Chemicals
399360010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 70491-05-3 | |
| 257.33 | |
| QSUXZIPXYDQFCX-SNVBAGLBSA-N | |
| 7014890 | |
| CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O |
| C13H23NO4 | |
| MFCD00133629 | |
| boc-d-chg-oh, boc-d-cyclohexylglycine, r-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-alpha-cyclohexyl-d-glycine, n-boc-2-cyclohexyl-d-glycine, 2r-n-tert-butoxycarbonyl-2-cyclohexylglycine, r-tert-butoxycarbonylamino-cyclohexyl-acetic acid, r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-2-tert-butoxycarbonylamino-2-cyclohexyl-acetic acid | |
| (2R)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid |
Specifications
| 70491-05-3 | |
| Glass bottle | |
| MFCD00133629 | |
| boc-d-chg-oh, boc-d-cyclohexylglycine, r-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-alpha-cyclohexyl-d-glycine, n-boc-2-cyclohexyl-d-glycine, 2r-n-tert-butoxycarbonyl-2-cyclohexylglycine, r-tert-butoxycarbonylamino-cyclohexyl-acetic acid, r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2r-2-tert-butoxycarbonylamino-2-cyclohexyl-acetic acid | |
| CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O | |
| 257.33 | |
| 257.33 | |
| BOC-D-α-Cyclohexylglycine |
| 98% | |
| C13H23NO4 | |
| 1g | |
| QSUXZIPXYDQFCX-SNVBAGLBSA-N | |
| (2R)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid | |
| 7014890 | |
| 98% |
RUO â Research Use Only