missing translation for 'onlineSavingsMsg'
Learn More

(Boc-amino)-PEG4-carboxylic Acid 98.0+%, TCI America™

$247.92 - $745.62

Chemical Identifiers

CAS 756525-91-4
Molecular Formula C16H31NO8
Molecular Weight (g/mol) 365.423
MDL Number MFCD07781254
InChI Key YEIYIPDFZMLJQH-UHFFFAOYSA-N
Synonym 15-[(tert-Butoxycarbonyl)amino]-4,7,10,13-tetraoxapentadecanoic Acid, 15-(Boc-amino)-4,7,10,13-tetraoxapentadecanoic Acid, 2,2-Dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azaicosan-20-oic Acid, Carboxy-PEG4-(Boc-amine)
PubChem CID 2756001
IUPAC Name 3-[2-[2-[2-[2-[(2-methylpropan-2-yl)oxycarbonylamino]ethoxy]ethoxy]ethoxy]ethoxy]propanoic acid
SMILES CC(C)(C)OC(=O)NCCOCCOCCOCCOCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B5665250MG
View Documents
TCI America
B5665250MG
250 mg
Each for $247.92
Only null left
 
B56651G
View Documents
TCI America
B56651G
1 g
Each for $745.62
Only null left
 
Description

Description

Specifications

Chemical Identifiers

756525-91-4
365.423
YEIYIPDFZMLJQH-UHFFFAOYSA-N
2756001
CC(C)(C)OC(=O)NCCOCCOCCOCCOCCC(=O)O
C16H31NO8
MFCD07781254
15-[(tert-Butoxycarbonyl)amino]-4,7,10,13-tetraoxapentadecanoic Acid, 15-(Boc-amino)-4,7,10,13-tetraoxapentadecanoic Acid, 2,2-Dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azaicosan-20-oic Acid, Carboxy-PEG4-(Boc-amine)
3-[2-[2-[2-[2-[(2-methylpropan-2-yl)oxycarbonylamino]ethoxy]ethoxy]ethoxy]ethoxy]propanoic acid

Specifications

756525-91-4
C16H31NO8
15-[(tert-Butoxycarbonyl)amino]-4,7,10,13-tetraoxapentadecanoic Acid, 15-(Boc-amino)-4,7,10,13-tetraoxapentadecanoic Acid, 2,2-Dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azaicosan-20-oic Acid, Carboxy-PEG4-(Boc-amine)
CC(C)(C)OC(=O)NCCOCCOCCOCCOCCC(=O)O
250 mg
2756001
≥98.0% (HPLC,T)
(Boc-amino)-PEG4-carboxylic Acid
Yellow
MFCD07781254
YEIYIPDFZMLJQH-UHFFFAOYSA-N
3-[2-[2-[2-[2-[(2-methylpropan-2-yl)oxycarbonylamino]ethoxy]ethoxy]ethoxy]ethoxy]propanoic acid
365.423
365.42
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer (-20 °C)

SDS
missing translation for 'documents'

missing translation for 'documents'