Learn More
(+/-)-Boc-alpha-phosphonoglycine trimethyl ester, 95%
CAS: 89524-98-1 | C10H20NO7P | 297.24 g/mol
$120.56 - $1057.42
Chemical Identifiers
| CAS | 89524-98-1 |
|---|---|
| Molecular Formula | C10H20NO7P |
| Molecular Weight (g/mol) | 297.24 |
| MDL Number | MFCD02178893 |
| InChI Key | LJHAPRKTPAREGO-ZETCQYMHSA-N |
| Synonym | methyl 2-tert-butoxycarbonyl amino-2-dimethoxyphosphoryl acetate, +/--boc-alpha-phosphonoglycine trimethyl ester, n-boc-2-phosphonoglycine trimethyl ester, methyl n-boc-2-dimethylphosphono glycinate, methyl 2-tert-butoxycarbonylamino-2-dimethoxyphosphoryl-acetate, boc-alpha-phosphonoglycine trimethyl ester, n-butoxycarbonyl trimethylphosphonoglycine, methyl 2-tert-butoxy carbonyl amino-2-dimethoxyphosphoryl acetate, boc-a-phosphonoglycine trimethyl ester, boc-?-phosphonoglycine trimethyl ester |
| PubChem CID | 397380 |
| IUPAC Name | methyl 2-dimethoxyphosphoryl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate |
| SMILES | COC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2542103
|
Thermo Scientific Chemicals
B2542103 |
1 g |
Each for $120.56
|
|
|||||
|
AAB2542106
|
Thermo Scientific Chemicals
B2542106 |
5 g |
Each for $354.82
|
|
|||||
|
AAB2542114
|
Thermo Scientific Chemicals
B2542114 |
25 g |
Each for $1,057.42
|
|
|||||
Description
It is used as Wittig-Horner reagent for preparing (Z)-Boc-protected dehydroamino acid derivatives and saturated, unnatural amino acids by subsequent asymmetric hydrogenation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as Wittig-Horner reagent for preparing (Z)-Boc-protected dehydroamino acid derivatives and saturated, unnatural amino acids by subsequent asymmetric hydrogenation.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Chemical Identifiers
| 89524-98-1 | |
| 297.24 | |
| LJHAPRKTPAREGO-ZETCQYMHSA-N | |
| 397380 | |
| COC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC |
| C10H20NO7P | |
| MFCD02178893 | |
| methyl 2-tert-butoxycarbonyl amino-2-dimethoxyphosphoryl acetate, +/--boc-alpha-phosphonoglycine trimethyl ester, n-boc-2-phosphonoglycine trimethyl ester, methyl n-boc-2-dimethylphosphono glycinate, methyl 2-tert-butoxycarbonylamino-2-dimethoxyphosphoryl-acetate, boc-alpha-phosphonoglycine trimethyl ester, n-butoxycarbonyl trimethylphosphonoglycine, methyl 2-tert-butoxy carbonyl amino-2-dimethoxyphosphoryl acetate, boc-a-phosphonoglycine trimethyl ester, boc-?-phosphonoglycine trimethyl ester | |
| methyl 2-dimethoxyphosphoryl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate |
Specifications
| 89524-98-1 | |
| C10H20NO7P | |
| 1 g | |
| Insoluble in water. | |
| COC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC | |
| 297.24 | |
| 297.24 | |
| (+/-)-Boc-alpha-phosphonoglycine trimethyl ester |
| 67°C to 71°C | |
| MFCD02178893 | |
| methyl 2-tert-butoxycarbonyl amino-2-dimethoxyphosphoryl acetate, +/--boc-alpha-phosphonoglycine trimethyl ester, n-boc-2-phosphonoglycine trimethyl ester, methyl n-boc-2-dimethylphosphono glycinate, methyl 2-tert-butoxycarbonylamino-2-dimethoxyphosphoryl-acetate, boc-alpha-phosphonoglycine trimethyl ester, n-butoxycarbonyl trimethylphosphonoglycine, methyl 2-tert-butoxy carbonyl amino-2-dimethoxyphosphoryl acetate, boc-a-phosphonoglycine trimethyl ester, boc-?-phosphonoglycine trimethyl ester | |
| LJHAPRKTPAREGO-ZETCQYMHSA-N | |
| methyl 2-dimethoxyphosphoryl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate | |
| 397380 | |
| 95% |
Safety and Handling
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only