missing translation for 'onlineSavingsMsg'
Learn More

BOC-2-piperidylmethanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 437520100

Quantity 10g

 View more versions of this product

Catalog No. AC437520100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

157634-00-9
215.29
n-boc-piperidine-2-methanol, tert-butyl 2-hydroxymethyl piperidine-1-carboxylate, 1-boc-2-piperidinemethanol, n-boc-2-piperidinemethanol, 2-hydroxymethyl-1-n-boc-piperidine, 1-boc-2-hydroxymethyl-piperidine, boc-2-piperidylmethanol, 1-boc-2-hydroxymethylpiperidine, n-boc-2-hydroxymethyl piperidine, 2-hydroxymethyl-piperidine-1-carboxylic acid tert-butyl ester
tert-butyl 2-(hydroxymethyl)piperidine-1-carboxylate
C11H21NO3
PZTAGFCBNDBBFZ-UHFFFAOYSA-N
2763853
CC(C)(C)OC(=O)N1CCCCC1CO

Specifications

BOC-2-Piperidylmethanol
Authentic
Glass bottle
10g
PZTAGFCBNDBBFZ-UHFFFAOYSA-N
tert-butyl 2-(hydroxymethyl)piperidine-1-carboxylate
2763853
97%
157634-00-9
96% min. (GC)
C11H21NO3
n-boc-piperidine-2-methanol, tert-butyl 2-hydroxymethyl piperidine-1-carboxylate, 1-boc-2-piperidinemethanol, n-boc-2-piperidinemethanol, 2-hydroxymethyl-1-n-boc-piperidine, 1-boc-2-hydroxymethyl-piperidine, boc-2-piperidylmethanol, 1-boc-2-hydroxymethylpiperidine, n-boc-2-hydroxymethyl piperidine, 2-hydroxymethyl-piperidine-1-carboxylic acid tert-butyl ester
CC(C)(C)OC(=O)N1CCCCC1CO
215.29
215.29
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions