missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BMP™61 Series Printer Ribbon - R6400 Resin, Black

Catalog No. 19150626
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19150626 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19150626 Supplier Brady™ Supplier No. M61R6410

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BMP™61 Series ribbon cartridges allow users to change ribbons in seconds.

  • One-color ribbon for printing a single color onto a label
  • R6400 Series Resin Formulation offers superior chemical and abrasion resistance when printed on the appropriate label material from Brady™
  • Recommended for use with materials designed to resist chemicals, such as B-492 Frozen Surface Polyester and B-481 Chemical Resistant Polyester
  • Automatic label formatting with smart cell technology and easy loading

Specifications

Color Black
For Use With (Equipment) BMP™61 Labeling System