missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BMP™61 Series Printer Ribbon - R4300 Wax/Resin, Black

Catalog No. 19150620
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19150620 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19150620 Supplier Brady™ Supplier No. M61R4310

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BMP™61 Series ribbon cartridges allow users to change ribbons in seconds.

  • One-color ribbon for printing a single color onto a label
  • R4300 Series Ribbons offers strong environmental resistance against smearing and chemicals when printed on the appropriate Brady™ label material
  • Recommended for printing on paper and films that have a matte or rough surface, such as B-427 Self-Laminating Vinyl and B-499 Nylon Cloth from Brady™
  • Automatic label formatting with smart cell technology and easy loading

Specifications

Product Type Printer Ribbon
Color Black
For Use With (Equipment) BMP61 Labeling System
Material Polyester
Print Technology Thermal Transfer
Length (English) 75 ft.
Width (English) 4.31 in.