MW 374.2 g/mol, Purity >98%. Chromogenic substrate for -galactosidase, used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates. Designed as an alternative to X-Gal (ab144388) in blue-white selection of recombinant bacterial colonies with the lac+ phenotype, producing a darker blue color.
The product is subject to the following: Abcam Restricted Use Statement