missing translation for 'onlineSavingsMsg'
Learn More

Blue Tetrazolium 95.0+%, TCI America™

Catalog No. B35811G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35811G 1 g
B35815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35811G Supplier TCI America Supplier No. B35811G

Chemical Identifiers

CAS 1871-22-3
Molecular Formula C40H36Cl2N8O2
Molecular Weight (g/mol) 731.68
MDL Number MFCD00040933
InChI Key RCEHREKDVGHYAM-UHFFFAOYSA-N
Synonym Ditetrazolium Chloride, Tetrazolium Blue
PubChem CID 9853362
ChEBI CHEBI:75198
IUPAC Name dihydrogen 2-[4'-(3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazol-2-yl)-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl]-3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazole dichloride
SMILES [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N1NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(N2NC(=NN2C2=CC=CC=C2)C2=CC=CC=C2)C(OC)=C1

Specifications

Melting Point 248°C
Formula Weight 727.65
Percent Purity ≥95.0% (T)
Color Yellow
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Blue Tetrazolium [for Biochemical Research]