Learn More
New England Biolab Ltd Blue Prestained Protein Standard, Broad Range (11-250 kDa) 150 gel lanes
Catalog No. NC1676285
Encompass
The Blue Prestained Protein Standard, Broad Range is a mixture of highly pure, recombinant, prestained proteins, covalently coupled with a blue chromophore, that resolves into 11 sharp bands when electrophoresed.
Catalog No. NC1676285 Supplier New England Biolab Ltd Supplier No. P7718S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Blue Prestained Protein Standard, Broad Range is a mixture of highly pure, recombinant, prestained proteins, covalently coupled with a blue chromophore, that resolves into 11 sharp bands when electrophoresed.