missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ BLS 1255 12 in. Double-Sided Laminate
Description
- This item is the laminate cartridge only (BLS1255 Laminating System sold separately)
- Industrial grade, double-sided laminate from Brady™ is constructed from tough polyester for durability even in harsh environments
- Significantly superior to typical polypropylene overlaminates in terms of outdoor performance and abrasion resistance
Specifications
Specifications
| Product Type | 12 in. double-sided polyester laminate cartridge |
| Color | Clear |
| For Use With (Equipment) | BLS1255 Laminating System |
| Material | Polyester |
| Width (English) | 12 in. |
| Sterility | Non-sterile |