missing translation for 'onlineSavingsMsg'
Learn More

Azure Biosystems Blot Washing Buffer

Catalog No. AC2113AZ
Change view
Click to view available options
Concentration:
10X
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
AC2113AZ 10X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC2113AZ Supplier Azure Biosystems Supplier No. AC2113

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Western blot washing solution, 10x concentrate, 500 mL

  • Wash buffers are an important part of the blotting process for removing unhybridized antibody and preventing background signal.
  • Blot washing buffer for use with any Western blotting system. Provided as a 10x concentrated solution
  • Size: 500 mL
  • NOTE: While this Blot Washing Buffer is compatible with NIR Fluorescent Western blots, for the best results with NIR fluorescence, we recommend the Azure Fluorescent Blot Washing Buffer.

Specifications

Chemical Name or Material Blot Washing Buffer
Concentration 10X
For Use With (Application) Western Blot
Physical Form Liquid