Learn More
Sol-M™ Blood Lancet
Catalog No. 361060270
Change view
Click to view available options
Needle Gauge:
28 G
30 G
33 G
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
361060270 30 G
361060271 28 G
361060283 33 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 361060270 Supplier Sol-M™ Supplier No. LAN001
Standard blood lancet used for capillary blood sampling.
  • Used for capillary blood sampling required in daily diabetes management programs
  • Tri-bevel cut cannula supports proper blood sample size
  • Needle is equipped with a protective cap for increased safety and easier placement
Specifications
Needle Gauge 30 G
Product Type Lancet