Learn More
Hardy Diagnostics™ Blood Agar Plate
This 5% sheep blood in tryptic soy agar plate is recommended for use as a general purpose growth media for the isolation, cultivation, and differentiation of a wide variety of microorganisms.
$29.06 - $29.06
Specifications
| Product Type | Agar |
|---|---|
| Sterility | Sterile |
| Test Type | Growth Promotion Testing |
| Catalog Number | Mfr. No. | No. per Pack | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. per Pack | Price | Quantity | |||||
|
23-001-791
|
Hardy Diagnostics™
A10* |
10 |
Pack of 10 Each for $29.06
|
|
|||||
Description
- Contains 5% sheep blood from a veterinary supervised, controlled herd of donor animals.
- Blood guaranteed to be from healthy antibiotic-free animals.
- Heavy duty reduces breaking during shiing
- Polystyrene Petri plates feature raised rings for stacking abity and prevents sliding
- Packaged in sleeves of 10 plates, in a 'breathable' bag prevents buildup of moisture
Specifications
| Agar | |
| Sterile | |
| Growth Promotion Testing |