missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hardy Diagnostics™ Blood Agar Plate
Description
- Contains 5% sheep blood from a veterinary supervised, controlled herd of donor animals.
- Blood guaranteed to be from healthy antibiotic-free animals.
- Heavy duty reduces breaking during shiing
- Polystyrene Petri plates feature raised rings for stacking abity and prevents sliding
- Packaged in sleeves of 10 plates, in a 'breathable' bag prevents buildup of moisture
Specifications
Specifications
| Certifications/Compliance | ISO 13485, ISO 17025 |
| Color | Cherry Red |
| Content And Storage | 2°C to 8°C |
| Format | Plate |
| Media Type | Blood Agar |
| Product Type | Agar Plate |
| Sterility | Sterile |
| Target Organism Class | Gram-positive Organisms |
| Test Type | Growth Promotion Testing |
| Fill Volume | 19 mL |
| Show More |