Learn More
StatLab Block Trimmer +
Description
- The edges and writing surface of the tissue cassette are de-waxed effortlessly by pressing the paraffin block onto the patterned heated work surface
- The Block Trimmer+ allows users to prepare tissue blocks with no risk of damage to the printed text or barcode on the tissue cassette
- An extra-large heated work surface allows multiple blocks to be pressed across the work surface at one time, significantly improving laboratory workflows
- Melted paraffin wax is then collected in a disposable collection pot
- Features: Small footprint, ultra-fast rapid warm-up, smart variable digital control remembers desired temperature at start-up, anti-microbial powder coat finish, and integrated miniature circuit breakers to increase safety and eliminate the need for replacement fuses
- Dimensions: 202 mm x 152 mm x 183.5 mm (D x W x H)
- Weight: 5 lb. (3 Kg)
Specifications
Specifications
| Product Type | Paraffin Block Trimmer |