missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Block Style Calorimeter

Block Style Calorimeter are used to determine the specific heat and heat capacity of different materials.

$76.70 - $416.47

Specifications

Grade 7 to Undergraduate
For Use With (Application) Specific Heat and Heat Capacity Determination
Includes 1 Calorimeter
Product Type Calorimeter (Block-style)
View More Specs

Products 4
Catalog Number Mfr. No. Material Price Quantity  
Catalog Number Mfr. No. Material Price Quantity  
S27678
View Documents
Eisco™
PH0435A
Aluminum N/A
Only null left
 
S27681
View Documents
Eisco™
PH0435D
Brass N/A
Only null left
 
S27680
View Documents
Eisco™
PH0435C
Copper N/A
Only null left
 
S27679
View Documents
Eisco™
PH0435B
Milled Steel N/A
Only null left
 
Description

Description

  • Made of high quality materials.
  • Each 1 kg aluminum block is drilled with two holes.
  • Large center hole has a diameter of 12.5 mm for a special immersion heater.
  • Small hole has a diameter of 7.5 mm for a thermometer or temperature sensor.
Specifications

Specifications

7 to Undergraduate
Specific Heat and Heat Capacity Determination
1 Calorimeter
Calorimeter (Block-style)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'